Petroleumbenzin, 35…60 °C

Lieferant: Thermo Scientific
Gefahr

Synonyms: BenzinPetrolether

40982.K2EA 102 CHF
40982.K2
Petroleumbenzin, 35…60 °C
Petroleumbenzin

Suitable for environmental and pesticide analysis

Siedepunkt: 35…60 °C
Dichte: 0,640
Flash-Pt: –49 °C (–57 °F)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00081849
CAS-Nummer: 64742-49-0
EINECS: 232-453-7
UN: 1268
ADR: 3,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR