Aluminium ≥99,5% (Metall-Basis), Pulver -100+325 mesh
Lieferant: Thermo Fisher Scientific
Gefahr
Synonyms: Aluminum
43022.18 43022.30 43022.A1
43022.18EA 110 CHF
43022.18 43022.30 43022.A1
Aluminium ≥99,5% (Metall-Basis), Pulver -100+325 mesh
Aluminium
Purity: 99.5% CAS No.: 7429-90-5 Physical Form: Powder Sensitivity: Moisture Sensitive Appearance: Silver-white Odour: Odourless UN No.: 1396 MDL No.: MFCD00134029
Formel: Al
MW: 26.98 g/mol
Siedepunkt: 2327
Melting Pt: 660
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00134029
CAS-Nummer: 7429-90-5
EINECS: 231-072-3
UN: 1396
ADR: 4.3 II
Merck Index: 13,00321