Aluminium ≥99,5% (Metall-Basis), Pulver -325 mesh
Lieferant: Thermo Fisher Scientific
Gefahr
Synonyms: Aluminum
11067.14 11067.30 11067.A1 11067.A7
11067.14EA 88.1 CHF
11067.14 11067.30 11067.A1 11067.A7
Aluminium ≥99,5% (Metall-Basis), Pulver -325 mesh
Aluminium
Purity: 99.5% CAS No.: 7429-90-5 Physical Form: APS 7-15 micron Sensitivity: Moisture Sensitive Odour: Odourless UN No.: 1396 MDL No.: MFCD00134029
Formel: Al
MW: 26.98 g/mol
Siedepunkt: 2327
Melting Pt: 660
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00134029
CAS-Nummer: 7429-90-5
EINECS: 231-072-3
UN: 1396
ADR: 4.3 II
Merck Index: 13,00321