Jars, wide mouth, with screw cap, Sterilin®
Lieferant: Thermo Fisher Scientific
215-2289EA 839 CHF
215-2289
Jars, wide mouth, with screw cap, Sterilin®
Probenbehälter Universalbehälter
PS, transparent with white urea plastic wadded cap.
  • Wide mouth enables bulk one piece specimens to be stored easily
  • Suitable for direct transfer of specimen from patient to jar
  • CE marked in accordance with the European Directive 98/79/EC
Multi use disposable jars. Ideal for liquid, solid, food and histology samples.