Protoporphyrin IX freie Säure ≥95% (durch HPLC)

Lieferant: Enzo Life Sciences

ALEXALX430041M300EA 109 CHF
ALEXALX430041M300 ALEXALX430041G001
Protoporphyrin IX freie Säure ≥95% (durch HPLC)
Protoporphyrin IX

Protoporphyrin IX free acid, as distinct from its zinc salt (which inhibits heme oxygenase), activates soluble guanylyl cyclase (sGC) by binding directly to the enzyme. A useful reagent in cases where the use of nitric oxide (NO) or nitric oxide donors is undesirable.


  • Guanylyl cyclase activator


Purple to dark brown solid.


Dissolve initially in 0.1M base (Tris base or NaOH), then add water miscible organic solvent (EtOH (10mg/ml), MeOH, DMSO (10mg/ml), DMF) until solution is 50/50 base/solvent. When a clear solution is obtained, it can be diluted into an aqueous medium and titrated or buffered to any pH>7. Use immediately.

MW: 562,66 g/mol
MDL Number: MFCD00151109
CAS-Nummer: 553-12-8
https://ch.vwr.com/stibo/web/std.lang.all/06/51/45390651.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR