Antigen retrieval reagent, pH 6 (10X)
Lieferant: Enzo Life Sciences
ALEXENZACC112-0100EA 176 CHF
ALEXENZACC112-0100
Antigen retrieval reagent, pH 6 (10X)
Protein-Extraktion Proteinextraktions-Reagenzien
Antigen Retrieval Reagent, pH 6 (10X) is used to recover antigens masked by fixation in cross linking fixatives such as formalin.