ApoStrand™ ELISA apoptosis detection kit
Lieferant: Enzo Life Sciences
ALEXBML-AK120-0001EA 747 CHF
ALEXBML-AK120-0001
ApoStrand™ ELISA apoptosis detection kit
Tests ELISAs
The ApoStrand™ ELISA is based on the sensitivity of DNA in apoptotic cells to formamide denaturation and the detection of the denatured DNA with an antibody to single-stranded DNA. It allows for the sensitive, specific and universal detection of apoptotic cells in a convenient 96 well microplate format. The ApoStrand™ ELISA can detect 500 apoptotic cells per well and gives a linear response in the range of 500 to 5000 cells. It will not give a positive signal for necrotic cells. The ApoStrand™ ELISA can be completed in 3-4 hours and is suitable for high-throughput screening.
Lieferumfang: Kit contains Antibody, ssDNA, Formamide, Blocking Solution, Peroxidase Substrate, Wash Buffer, Fixative and Sterile 96-well plate.