p53 (Human) ELISA Kit
Lieferant: Enzo Life Sciences
ALEXALX850057KI01EA 0 CHF
ALEXALX850057KI01
p53 (Human) ELISA Kit
Tests ELISAs
For the quantitative determination of human p53 in cell culture supernatants, human serum, plasma and other biological fluids.
  • Assay Time: 3 hours
p53 plays an important role in anticancer and works through many mechanisms to control DNA repair. When DNA has sustained damage, p53 is capable of activating DNA repair and can also initiate apoptosis when DNA damage is irreparable.