Anti-CD23 Mouse Monoclonal Antibody (Biotin) [clone: 9p25]
Lieferant: Axxora
ALEXENZABS2470100EA 585 CHF
ALEXENZABS2470100 ENZOENZABS2470100
Anti-CD23 Mouse Monoclonal Antibody (Biotin) [clone: 9p25]
Antikörper
Recommended Applications: Flow Cytometry
Species reactivity: Human
Type: Primary
Antigen: CD23
Clonality: Monoclonal
Clone: 9p25
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human