Anti-CD39 Mouse Monoclonal Antibody [clone: AC2.5]

Lieferant: Axxora
ENZOENZABS2330100EA 738 CHF
ENZOENZABS2330100 ALEXENZABS2330100
Anti-CD39 Mouse Monoclonal Antibody [clone: AC2.5]
Antikörper
Recommended Applications: Flow Cytometry, IHC (FS), IP

Species reactivity: Human

Type: Primary
Antigen: CD39
Clonality: Monoclonal
Clone: AC2.5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR