Anti-GRP78 Rabbit Polyclonal Antibody

Lieferant: Axxora
ALEXALX210137C100EA 856 CHF
ALEXALX210137C100
Anti-GRP78 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Grp78
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR