Anti-CK7 Mouse Monoclonal Antibody [clone: 4F5]

Lieferant: ENZO LIFE SCIENCES AG
ENZOBMLCA11390100EA 249 CHF
ENZOBMLCA11390100 ALEXBMLCA11390100
Anti-CK7 Mouse Monoclonal Antibody [clone: 4F5]
Antikörper
Host: Mouse, Isotype: IgG1

  • For Immunocytochemistry, Western Blot

Recognizes human, mouse, rat, pig cytokeratin proteins 7, 17 and 19 respectively. Detects bands of 54 kDa, 46 kDa and 40 kDa in Western Blot.

Type: Primary
Antigen: CK7
Clonality: Monoclonal
Clone: 4F5
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR