Anti-B2M Mouse Monoclonal Antibody
Lieferant: Enzo Life Sciences
ALEXALX804482C100EA 263 CHF
ALEXALX804482C100
Anti-B2M Mouse Monoclonal Antibody
Antikörper
Host: Mouse, Isotype: IgG2a
  • For ELISA, Flow Cytometry, Immunoprecipitation, Western Blot
Recognizes human beta2-microglobulin associated with cell-surface MHC Class I molecules and other membrane antigens as well as soluble human beta2-microglobulin. Does not cross-react with mouse, bovine, rabbit, dog or chicken beta2-microglobulin.
Type: Primary
Antigen: B2M
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human