Anti-DLC1 Rat Monoclonal Antibody
Lieferant: Enzo Life Sciences
ALEXALX804340C100EA 516 CHF
ALEXALX804340C100
Anti-DLC1 Rat Monoclonal Antibody
Antikörper
Host: Rat, Isotype: IgM
  • For Western Blot
Recognizes human and mouse DLC1. Does not cross-react with DLC2. For detection of both DLC1 and DLC2 see MAb to DLC1 and DLC2 (11F7) (Prod. No. ALX-804-341).
Type: Primary
Antigen: DLC1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rat
Isotype: IgM
Reactivity: Human