Anti-SOD2 Mouse Monoclonal Antibody

Lieferant: Enzo Life Sciences
ALEXALX804265C100EA 444 CHF
ALEXALX804265C100
Anti-SOD2 Mouse Monoclonal Antibody
Antikörper
Host: Mouse, Isotype: IgG1

  • For Immunohistochemistry, Western Blot

Recognizes human Mn SOD. Cross-reacts with rat and rabbit Mn SOD.

Type: Primary
Antigen: SOD2
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR