Anti-I kappa B alpha Mouse Monoclonal Antibody

Lieferant: Enzo Life Sciences
ALEXALX804209C100EA 545 CHF
ALEXALX804209C100
Anti-I kappa B alpha Mouse Monoclonal Antibody
Antikörper
Host: Mouse, Isotype: IgG1

  • For Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes both phosphorylated and non-phosphorylated forms of human and mouse IkappaBalpha. Detects a band of ~40kDa by Western blot.

Type: Primary
Antigen: IKAPPABALPHA
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR