Anti-MRP3 Mouse Monoclonal Antibody

Lieferant: Enzo Life Sciences
ALEXALX801019C250EA 1130 CHF
ALEXALX801019C250
Anti-MRP3 Mouse Monoclonal Antibody
Antikörper
Host: Mouse, Isotype: IgG1

  • For Immunocytochemistry, Immunohistochemistry, Western Blot

Recognizes an internal epitope of human MRP3. Does not cross-react with human P-glycoprotein, MDR3 P-glycoprotein, MRP1, MRP2 (cMOAT) or MRP5. Does not cross-react with mouse, rat or guinea pig MRP2.

Type: Primary
Antigen: MRP3
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR