Anti-P-Glycoprotein Mouse Monoclonal Antibody
Lieferant: Enzo Life Sciences
ALEXALX801003C100EA 1560 CHF
ALEXALX801003C100
Anti-P-Glycoprotein Mouse Monoclonal Antibody
Antikörper
Host: Mouse, Isotype: IgG2a
  • For Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Recognizes a gene-specific, internal cellular epitope present only on the human MDR1 isoform of P-glycoprotein. Does not cross-react with human MDR3 P-glycoprotein.
Type: Primary
Antigen: P-GLYCOPROTEIN
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human