Anti-CCR7 Goat Polyclonal Antibody
Lieferant: Enzo Life Sciences
ALEXALX210844C100EA 568 CHF
ALEXALX210844C100
Anti-CCR7 Goat Polyclonal Antibody
Antikörper
Host: Goat
  • For Immunocytochemistry, Immunohistochemistry, Western Blot
Recognizes human and mouse CCR7.
Type: Primary
Antigen: CCR7
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human