Anti-CCR5 Goat Polyclonal Antibody

Lieferant: Enzo Life Sciences
ALEXALX210823C200EA 472 CHF
ALEXALX210823C200
Anti-CCR5 Goat Polyclonal Antibody
Antikörper
Host: Goat

  • For ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot

Recognizes human CCR5.

Type: Primary
Antigen: CCR5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR