Anti-CASP11 Rabbit Polyclonal Antibody
Lieferant: Enzo Life Sciences
ALEXALX210818C100EA 450 CHF
ALEXALX210818C100
Anti-CASP11 Rabbit Polyclonal Antibody
Antikörper
Host: Rabbit
  • For Western Blot
Recognizes mouse and rat caspase-11. Detects full length caspase-11 and cleavage fragments by Western blot.
Type: Primary
Antigen: CASP11
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse