Anti-P110D Rabbit Polyclonal Antibody

Lieferant: Enzo Life Sciences
ALEXALX210747C200EA 684 CHF
ALEXALX210747C200
Anti-P110D Rabbit Polyclonal Antibody
Antikörper
Host: Rabbit

  • For Immunocytochemistry, Immunohistochemistry, Western Blot

Recognizes human PI(3)K p110delta. Detects a band of ~110kDa by Western blot.

Type: Primary
Antigen: P110D
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR