Anti-FURIN Rabbit Polyclonal Antibody

Lieferant: Enzo Life Sciences
ALEXALX210134R100EA 938 CHF
ALEXALX210134R100
Anti-FURIN Rabbit Polyclonal Antibody
Antikörper
Host: Rabbit

  • For Immunocytochemistry, Immunoprecipitation, Western Blot

Recognizes mouse and dog furin convertase and overexpressed human furin convertase. Detects a band of ~100kDa by Western blot. Does not detect endogenous furin from BSC-40, HeLa, J774A-1 BPAEC or CHO cells. Does not cross-react with rat furin convertase.

Type: Primary
Antigen: FURIN
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR