Electrolysis apparatus according to Hoffmann
Lieferant: NEUBERT VOLUME GLASSWAERE
0500-10-40 0500-10-45 0500-10-30 0500-10-33
NEUB0500-10-40EA 496 CHF
NEUB0500-10-40 NEUB0500-10-45 NEUB0500-10-30 NEUB0500-10-33
Electrolysis apparatus according to Hoffmann
Lehrmittel Chemie-Lehrmittel Electrochemistry Learning Activities
Electrolysis apparatus according to Hoffmann, made of borosilicate glass 3.3.
  • Connecting piece for more stability
  • Glass tap NS 12
  • Graduated from 0 - 60 ml with 0,2 ml intervals
Order Now
TECHNISCHE DATEN
Environmentally Preferable