Anti-TNFRSF13C Rat Monoclonal Antibody (Biotin) [clone: 9B9]

Lieferant: ProSci
PRSI36-143EA 712 CHF
PRSI36-143
Anti-TNFRSF13C Rat Monoclonal Antibody (Biotin) [clone: 9B9]
Antikörper
Three receptors for BAFF were recently identified and designated TACI, BCMA and BAFF-R. BAFF-R signaling is essential for B cell development, survival and humoral immune responses.

Functional Application: Depletion of B cells in vivo. Optimal conditions must be determined individually for each application.

Type: Primary
Antigen: TNFRSF13C
Clonality: Monoclonal
Clone: 9B9
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR