Anti-TNFRSF18 Rat Monoclonal Antibody [clone: AIT 158D]
Lieferant: ProSci
PRSI36-016EA 528 CHF
PRSI36-016
Anti-TNFRSF18 Rat Monoclonal Antibody [clone: AIT 158D]
Antikörper
GITR is the receptor for TNFSF18. It is involved in interactions between activated T-lymphocytes and endothelial cells and in the regulation of T-cell receptor-mediated cell death. Mediates NF-kappaB activation via the TRAF2/NIK pathway.
ELISA: (direct and indirect: 1:5,000-1:10,000). Flow Cytometry: (1:1,000). Optimal conditions must be determined individually for each application.
Type: Primary
Antigen: TNFRSF18
Clonality: Monoclonal
Clone: AIT 158D
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Human