Labels, tags, energy source, valve
Lieferant: Brady
BRDY138440EA 116 CHF
BRDY138440 BRDY138792 BRDY138791 BRDY138790 BRDY138439 BRDY138801 BRDY138796 BRDY138443 BRDY138795 BRDY138442 BRDY138794 BRDY138793 BRDY138441 BRDY138800 BRDY138789 BRDY138788 BRDY138799 BRDY138787 BRDY138798 BRDY138797
Labels, tags, energy source, valve
Etiketten
These valve energy source tags are constructed of PP with round corners with one hole.
  • Permanent acrylic adhesive
  • Helps the lockout process quicker and easier for higher safety
Used to mark your facility's points of control of control for energy sources.
Colour: Black on white
Zertifizierungen: ATEX
Verpackung: 25 tags per pack.