[1,1'-Bis(diphenylphosphino)ferrocen]palladium(II)chlorid Pd 13,5-15,5%

Achtung

ALFA46503.03EA 99.8 CHF
ALFA46503.03
[1,1'-Bis(diphenylphosphino)ferrocen]palladium(II)chlorid Pd 13,5-15,5%
[1,1'-Bis(diphenylphosphino)ferrocen]palladium(II)chlorid
Formel: C₃₄H₂₈Cl₂P₂FePd
MW: 731,70 g/mol
Melting Pt: 266…283 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00015757
CAS-Nummer: 72287-26-4
EINECS: 460-040-3

Order Now

Spezifikation Testergebnisse

Formula C34H28Cl2FeP2Pd
Formula weight 731,72
Form Solid
Melting point 266 - 283°
Solubility Soluble in dichloromethane
Assay (unspecified) Pd 13,5 - 15,5%
CAS number 72287-26-4
MDL number MFCD00015757


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR