Natriumchlorid Ph. Eur., Macron Fine Chemicals™
Lieferant: Avantor Performance Materials
P135-38EA 283 CHF
P135-38 P135-33
Natriumchlorid Ph. Eur., Macron Fine Chemicals™
Natriumchlorid
Formel: NaCl
MW: 58,44 g/mol
MDL Number: MFCD00003477
CAS-Nummer: 7647-14-5