Detergents, InSpec™ N10
Lieferant: Dycem
InSpec™ N10
DYCEINSPECN10EA 563 CHF
DYCEINSPECN10
Detergents, InSpec™ N10
Reinigungsmittel Waschmittel und Oberflächenreiniger
Inspec N10 is a powerful non sterile solution of mildly alkaline detergent specifically used for cleanroom walls, floors, ceilings and work surfaces. It is most effective at removing general process derived residue and soiling.
  • Rapid action alkaline detergent
  • Suitable for manual cleaning
  • Defined 24 month shelf life
This is also available as a pre-diluted detergent, made with WFI quality water.Available in sterile or non sterile preparations.
Zertifizierungen: Manufactured to GMP standards in an ISO 6 cleanroom, filled and capped in an ISO class 5 environment.
Verpackung: Solutions are filled through a 0,2 micron filter into final packaging.