APLIA0903
Lieferant: PanReac AppliChem
A0903.0005EA 520 CHF
A0903.0005 A0903.0001
APLIA0903
It binds to protein kinase C and leads to a very strong activation of this enzyme. Physiological effects are very diverse.
TPA is soluble in DMSO or ethanol. The crystalline substance as well as the stock solution should be stored in the dark at −20 °C (stock solution 1 mg/ml; working concentration 100 ng/ml). Long term storage should be at −70 °C.