Bubble trap
Lieferant: Diba Industries
OMNI006BTEA 438 CHF
OMNI006BT
Bubble trap
Vakuumfallen
The Omnifit® labware bubble trap is made of PEEK. It effectively removes bubbles from aqueous solutions. The units are connected in-line, downstream of a pump, using 28 UNF threaded fittings (6,35 mm), such as Omni-Lok™.
  • Up to 6 ml/min flow rate
  • Quick, in-line connection with minimal maintenance
When a fluid containing bubbles flows through the unit, aqueous fluid is retained while bubbles are forced through a micro-porous, hydrophobic membrane (10 µm PTFE). The membrane function depends on its hydrophobicity, therefore the units are only suitable for use in aqueous systems and NOT with organic solvents.