Connecting adapters, for rotary evaporators to vials
Lieferant: CHEMGLASS
CG-1318-20 CG-1318-12 CG-1318-11 CG-1318-41 CG-1318-25 CG-1318-10 CG-1318-21 CG-1318-42 CG-1318-24 CG-1318-40 CG-1318-22 CG-1318-23
CGLACG-1318-20EA 17.74 CHF
CGLACG-1318-20 CGLACG-1318-12 CGLACG-1318-11 CGLACG-1318-41 CGLACG-1318-25 CGLACG-1318-10 CGLACG-1318-21 CGLACG-1318-42 CGLACG-1318-24 CGLACG-1318-40 CGLACG-1318-22 CGLACG-1318-23
Connecting adapters, for rotary evaporators to vials
Adapter Adapter, Normschliff mit Schlaucholiven
These adapters permit the connection of standard laboratory dram sample vials directly to the vapor tube or a bump trap of a rotary evaporator.
  • Very good chemical resistance
  • Very good temperature resistance
With a top outer joint of the listed size and a 20 to 400 G.P.I. thread at the bottom. A PTFE lined, polypropylene double-threaded connector attaches the vial to the adapter.
Bestellinformation: The glass adapter and double-threaded connections can be ordered separately.
Lieferumfang: Complete kits includes glass adapter and the 6 sizes of the double threaded adapter (packaged in a divided plastic box).