Glove bag
Lieferant: Glas-Col
112-9990EA 438 CHF
112-9990
Glove bag
Handschuhboxen und -beutel
PE glove bag with inflatable glove chamber, Handy-Lok closure, turned in gloves and one equipment opening.
  • Greater flexibility than a rigid glove box
  • Excels for working with air sensitive or moisture sensitive materials
  • Economical enough to use once and discard or durable enough to use over and over
  • Can be purged quickly and with less gas
Lieferumfang: Supplied with one pair of cotton gloves.