Anti-MCAM Mouse Monoclonal Antibody (CF568) [clone: MCAM/1101]

Lieferant: Biotium
BNC681101-500 BNC681101-100
BNC681101-500EA 671 CHF
BNC681101-500 BNC681101-100
Anti-MCAM Mouse Monoclonal Antibody (CF568) [clone: MCAM/1101]
Antikörper
The human Mel-CAM gene maps to chromosome 11q23 and encodes a trans-membrane glycoprotein, also designated MCAM, MUC 18 or CD146, that belongs to the immunoglobulin superfamily and functions as a Ca2 -independent cell adhesion molecule. Mel-CAM expression is restricted to advanced primary and metastatic melanomas and to cell lines of the neuroectodermal lineage, but not normal melanocytes. Mel-CAM is found on 80% of advanced primary human melanomas and correlates well with development of metastatic disease.

Type: Primary
Antigen: MCAM
Clonality: Monoclonal
Clone: MCAM/1101
Conjugation: CF568
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR