Anti-PDE6D Rabbit Polyclonal Antibody

Lieferant: Novus Biologicals
NOVUNBP1-32730EA 772 CHF
NOVUNBP1-32730
Anti-PDE6D Rabbit Polyclonal Antibody
Antikörper
Rabbit Polyclonal PDE6D Antibody. Tested Applications: Western Blot. Tested Reactivity: Human.

Type: Primary
Antigen: PDE6D
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR