Anti-GPR49 Rabbit Polyclonal Antibody
Lieferant: Novus Biologicals
NBP1-28904 NBP1-28904SS
NOVUNBP1-28904EA 837 CHF
NOVUNBP1-28904 NOVUNBP1-28904SS
Anti-GPR49 Rabbit Polyclonal Antibody
Antikörper
Rabbit Polyclonal GPR49/LGR5 Antibody. Tested Applications: Immunohistochemistry, Immunohistochemistry-Paraffin, ICC/IF (-). Tested Reactivity: Human.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:200
Type: Primary
Antigen: GPR49/LGR5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human