Anti-C3 Rabbit Polyclonal Antibody

Lieferant: Novus Biologicals
NOVUNBP1-32080EA 765 CHF
NOVUNBP1-32080
Anti-C3 Rabbit Polyclonal Antibody
Antikörper
Rabbit Polyclonal Complement C3 Antibody. Tested Applications: Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human, Mouse.

  • Recommended Dilutions: Immunohistochemistry (Parafin): 1:100-1:1000

Type: Primary
Antigen: C3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR