Anti-CD31 Rabbit Polyclonal Antibody
Lieferant: Novus Biologicals
NBP1-71663 NBP1-71663SS
NOVUNBP1-71663EA 677 CHF
NOVUNBP1-71663 NOVUNBP1-71663SS
Anti-CD31 Rabbit Polyclonal Antibody
Antikörper
Rabbit Polyclonal CD31/PECAM1 Antibody. Tested Applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human, Mouse.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:50
Type: Primary
Antigen: CD31
Clonality: Polyclonal
Clone: 0
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now