Anti-KI67 Rabbit Monoclonal Antibody

Lieferant: Novus Biologicals
NOVUNB600-1252EA 812 CHF
NOVUNB600-1252
Anti-KI67 Rabbit Monoclonal Antibody
Antikörper
Rabbit Monoclonal Ki67 Antibody (SP6). Tested Applications: Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human, Rat.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:100

Type: Primary
Antigen: KI67
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR