Anti-MYADM Mouse Monoclonal Antibody (CF640R) [clone: MYADM/972]
Lieferant: Biotium
BNC400972-500 BNC400972-100
BNC400972-500EA 671 CHF
BNC400972-500 BNC400972-100
Anti-MYADM Mouse Monoclonal Antibody (CF640R) [clone: MYADM/972]
Antikörper
Recognizes a myeloid associated differentiation antigen in the cytoplasm of mature granulocytes. It shows no reactivity with any other cell type in human tissues. Markers of myeloid cells are useful in the identification of different levels of cellular differentiation. It reacts with early precursor and mature forms of human and monkey myeloid cells. This MAb is useful for the detection of myeloid leukemias and granulocytic sarcomas. It can be used as a marker of granulocytes in normal tissues or inflammatory processes.
Type: Primary
Antigen: MYADM
Clonality: Monoclonal
Clone: MYADM/972
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Macaque