Anti-HA Epitope Tag Goat Polyclonal Antibody

Lieferant: Novus Biologicals
NOVUNB600-362EA 665 CHF
NOVUNB600-362
Anti-HA Epitope Tag Goat Polyclonal Antibody
Antikörper
Goat Polyclonal HA Epitope Tag Antibody. Tested Applications: Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation. Tested Reactivity: Non-species specific.

Type: Primary
Antigen: HA Epitope Tag
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Non-species specific
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR