Anti-ABO Mouse Monoclonal Antibody (Purified) [clone: 19-OLE]
Lieferant: Biotium
BNUB0947-500 BNUB0947-100
BNUB0947-500EA 613 CHF
BNUB0947-500 BNUB0947-100
Anti-ABO Mouse Monoclonal Antibody (Purified) [clone: 19-OLE]
Antikörper
Recognizes the blood group H type 2 antigens, trisaccharide Fucα1-2Galβ1-4GlcNAcβ1 of human origin. This protein is the basis of the ABO blood group system. The histo-blood group ABO involves three carbohydrate antigens: A, B, and H. A, B, and AB individuals express a glycosyltransferase activity that converts the H antigen to the A antigen (by addition of UDP-GalNAc) or to the B antigen (by addition of UDP-Gal), whereas O individuals lack such activity. It is expressed on endothelial cells, epithelial cells and granulocytes. Increased expression of this antigen has been observed on some tumor tissues such as gastric carcinomas, urothelial carcinomas, and colon carcinomas.
Type: Primary
Antigen: ABO
Clonality: Monoclonal
Clone: 19-OLE
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgM kappa
Reactivity: Human