Anti-CD40 Rat Monoclonal Antibody [clone: FGK45]

Lieferant: ADIPOGEN
AG20B0034BC100 AG20B0034C100
ADIPAG20B0034BC100EA 440 CHF
ADIPAG20B0034BC100 ADIPAG20B0034C100
Anti-CD40 Rat Monoclonal Antibody [clone: FGK45]
Antikörper
Type:
Antigen: CD40
Clonality: Monoclonal
Clone: FGK45
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR