Anti-NMNAT2 Mouse Monoclonal Antibody [clone: Nady-1]

Lieferant: AdipoGen Life Sciences
AG35B00082050 AG20A0086C050
ADIPAG35B00082050EA 350 CHF
ADIPAG35B00082050 ADIPAG20A0086C050
Anti-NMNAT2 Mouse Monoclonal Antibody [clone: Nady-1]
Antikörper
Type:
Antigen: NMNAT2
Clonality: Monoclonal
Clone: Nady-1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR