Anti-NQO1 Mouse Monoclonal Antibody [clone: Skiny-1]

Lieferant: AdipoGen Life Sciences
AG20A0085C050 AG20A0085C100
ADIPAG20A0085C050EA 110 CHF
ADIPAG20A0085C050 ADIPAG20A0085C100
Anti-NQO1 Mouse Monoclonal Antibody [clone: Skiny-1]
Antikörper
Type:
Antigen: NQO1
Clonality: Monoclonal
Clone: Skiny-1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR