Anti-Sirtuin Mouse Monoclonal Antibody [clone: S5R37-3]

Lieferant: AdipoGen Life Sciences
AG20A0083C050 AG20A0083C100
ADIPAG20A0083C050EA 300 CHF
ADIPAG20A0083C050 ADIPAG20A0083C100
Anti-Sirtuin Mouse Monoclonal Antibody [clone: S5R37-3]
Antikörper
Type:
Antigen: Sirtuin
Clonality: Monoclonal
Clone: S5R37-3
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR