Anti-Sirtuin Mouse Monoclonal Antibody [clone: SR119-1AG]

Lieferant: AdipoGen Life Sciences
AG20A0080C050 AG20A0080C100
ADIPAG20A0080C050EA 110 CHF
ADIPAG20A0080C050 ADIPAG20A0080C100
Anti-Sirtuin Mouse Monoclonal Antibody [clone: SR119-1AG]
Antikörper
Type:
Antigen: Sirtuin
Clonality: Monoclonal
Clone: SR119-1AG
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR