Anti-IL17A Mouse Monoclonal Antibody [clone: 17-142F]

Lieferant: AdipoGen Life Sciences
AG20A0070C050 AG20A0070C100
ADIPAG20A0070C050EA 300 CHF
ADIPAG20A0070C050 ADIPAG20A0070C100
Anti-IL17A Mouse Monoclonal Antibody [clone: 17-142F]
Antikörper
Type:
Antigen: IL17A
Clonality: Monoclonal
Clone: 17-142F
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR