Anti-DLK1 Rat Monoclonal Antibody [clone: PF105B]

Lieferant: ADIPOGEN
AG20A0055C100 AG20A0056C050
ADIPAG20A0055C100EA 190 CHF
ADIPAG20A0055C100 ADIPAG20A0056C050
Anti-DLK1 Rat Monoclonal Antibody [clone: PF105B]
Antikörper
Type:
Antigen: DLK1
Clonality: Monoclonal
Clone: PF105B
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR